Learn More
Raybiotech Inc C-Series Human Apoptosis Signaling Pathway Array C1.
Catalog No. 501948481
Encompass_Preferred
Detects 19 phosphorylated or cleaved human Apoptotic Factors. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Akt (P-Ser473) (PKB (P-Ser473)), ATM (P-Ser1981), BAD (P-Ser112), Caspase-3 (Cleaved Asp175), Caspase-7 (Cleaved Asp198), CHK1 (P-Ser296), CHK2 (P-Thr68), eIF-2a (P-Ser52), ERK1 (P-T202/Y204)/ERK2 (P-Y185/Y187), HSP27 (P-Ser82), IKBa (P-Ser32), JNK (P-Thr183/Tyr185), NFKBP65 (P-Ser536), PARP1 (Cleaved Asp214/Gly215), p27 (P-Thr198), P53 (P-Ser15), SMAD2 (Ser245/250/255), TAK1 (P-Ser412)
Catalog No. 50-194-8481 Supplier Raybiotech Inc Supplier No. AAHAPOSIG14
Detects 19 phosphorylated or cleaved human Apoptotic Factors. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Akt (P-Ser473) (PKB (P-Ser473)), ATM (P-Ser1981), BAD (P-Ser112), Caspase-3 (Cleaved Asp175), Caspase-7 (Cleaved Asp198), CHK1 (P-Ser296), CHK2 (P-Thr68), eIF-2a (P-Ser52), ERK1 (P-T202/Y204)/ERK2 (P-Y185/Y187), HSP27 (P-Ser82), IKBa (P-Ser32), JNK (P-Thr183/Tyr185), NFKBP65 (P-Ser536), PARP1 (Cleaved Asp214/Gly215), p27 (P-Thr198), P53 (P-Ser15), SMAD2 (Ser245/250/255), TAK1 (P-Ser412)