Learn More
Raybiotech Inc G-Series Human IL-1 Family Cytokine Array 1 Kit.
Catalog No. 501950405
Encompass_Preferred
Semi-quantitative measurement of 11 human IL-1 family cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 RA (IL-1 F3), IL-18, IL-33 (IL-1 F11), IL-36 alpha, IL-36 beta, IL-36 gamma, IL-36 RA, IL-37, IL-38
Catalog No. 50-195-0405 Supplier Raybiotech Inc Supplier No. GSHIL1F12
Semi-quantitative measurement of 11 human IL-1 family cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 RA (IL-1 F3), IL-18, IL-33 (IL-1 F11), IL-36 alpha, IL-36 beta, IL-36 gamma, IL-36 RA, IL-37, IL-38