Learn More
Raybiotech Inc G-Series Human Periodontal Disease Array 1 Kit.
Catalog No. 501950417
Encompass_Preferred
Semi-quantitative measurement of 20 human periodontal disease associated cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: CRP (C-Reactive Protein), IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-10, IL-12 p70, IL-17A, IL-2, IL-4, IL-6, IL-8 (CXCL8), MIP-1 alpha (CCL3), MMP-13, MMP-9, Osteoactivin (GPNMB), Osteopontin (SPP1), Osteoprotegerin (TNFRSF11B), RANK (TNFRSF11A), TGF beta 1, TNF alpha
Catalog No. 50-195-0417 Supplier Raybiotech Inc Supplier No. GSHPDD12
Semi-quantitative measurement of 20 human periodontal disease associated cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: CRP (C-Reactive Protein), IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-10, IL-12 p70, IL-17A, IL-2, IL-4, IL-6, IL-8 (CXCL8), MIP-1 alpha (CCL3), MMP-13, MMP-9, Osteoactivin (GPNMB), Osteopontin (SPP1), Osteoprotegerin (TNFRSF11B), RANK (TNFRSF11A), TGF beta 1, TNF alpha