Learn More
Raybiotech Inc G-Series Human Serum Abundant Protein Array 1 Kit.
Supplier:  Raybiotech Inc GSHSAP11
Encompass_Preferred
Semi-quantitative measurement of 31 Human Serum Abundant serum proteins. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: ALCAM (CD166), Angiotensinogen (SERPINA8), ANGPTL3, Cathepsin B (Pro-Cathepsin B), Chemerin, CHI3L1, Cystatin C, Decorin, DKK-3, ErbB3, GASP-1 (WFIKKNRP), ICAM-1 (CD54), IL-1 R3 (IL-1 R Acp), IL-6 R, LAP/TGF beta 1, MIP-1 delta (CCL15), MSP alpha/beta, NAP-2 (PPBP/CXCL7), NOV (CCN3), PARC (CCL18), P-Cadherin, PDGF-AA, PDGF-BB, PGRPs, RANTES (CCL5), SCF R (CD117/c-kit), Siglec-5 (CD170), TIMP-1, TIMP-2, Trappin-2, VCAM-1 (CD106)
Catalog No. 50-195-0420