Learn More
Raybiotech Inc G-Series Human Immune Response Array 1 Kit.
Supplier:  Raybiotech Inc GSHIMR11
Encompass_Preferred
Semi-quantitative measurement of 40 human immune response associated cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: CD14, CD163, CD40 (TNFRSF5), CRP (C-Reactive Protein), E-Selectin, Fas (TNFRSF6/Apo-1), Fas Ligand (TNFSF6), GCSF, ICAM-1 (CD54), IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 R4 (ST2), IL-10, IL-12 p70, IL-13, IL-18, IL-2, IL-2 R alpha, IL-4, IL-6, IL-8 (CXCL8), Lipocalin-2 (NGAL), MCP-1 (CCL2), MCP-2 (CCL8), MIF, MIP-1 alpha (CCL3), MIP-1 beta (CCL4), Osteopontin (SPP1), PAI-1, Platelet Factor 4 (CXCL4), Procalcitonin, RAGE, Resistin, Thrombomodulin, TNF alpha, TREM-1, Troponin I, uPAR, VCAM-1 (CD106), VEGF-A
Catalog No. 50-195-0406