Learn More
Raybiotech Inc C-Series Human Cancer Discovery Antibody Array 3 Kit.
Supplier:  Raybiotech Inc AAHCAN34
Encompass_Preferred
Detects 30 cytokines related to cancer biology. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Alpha-fetoprotein, Beta-2 Microglobulin, bFGF, CA125, CA19-9, CEA, CRP (C-Reactive Protein), DKK-1, ErbB2, Ferritin, FSH, Galectin-3, GDF-15, HGF, ICAM-1 (CD54), IL-8 (CXCL8), MIF, MMP-2, NSE, Osteoprotegerin (TNFRSF11B), Osteopontin (SPP1), PAI-1, PSA-free, RANTES (CCL5), SCF, TGF beta 1, Thrombospondin 1, TIMP-1, uPA, VEGF-A
Catalog No. 50-194-8522