Learn More
Raybiotech Inc G-Series Bovine Cytokine Array 3 Kit.
Supplier:  Raybiotech Inc GSHSAP42
Encompass_Preferred
Semi-Quantitative measurement of 40 Human Serum Abundant serum proteins. Suitable for all liquid sample types.
Catalog No. 50-195-0427