Learn More
Raybiotech Inc G-Series Non-Human Primate Cytokine Array 1 Kit.
Catalog No. 501950474
Encompass_Preferred
Semi-quantitative measurement of 10 Non-Human Primate (NHP) cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: GM-CSF, IFN-gamma, IL-1 beta (IL-1 F2), IL-4, IL-5, IL-6, IL-12 p70, IL-15, IL-16, TNF alpha
Catalog No. 50-195-0474 Supplier Raybiotech Inc Supplier No. GSNCYT11
Semi-quantitative measurement of 10 Non-Human Primate (NHP) cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: GM-CSF, IFN-gamma, IL-1 beta (IL-1 F2), IL-4, IL-5, IL-6, IL-12 p70, IL-15, IL-16, TNF alpha