Learn More
Raybiotech Inc Quantibody Human Immune Checkpoint Molecule Array 1 Kit.
Supplier:  Raybiotech Inc QAHICM12
Encompass_Preferred
Detects 10 Human Immune Checkpoint Molecules. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: CD80 (B7-1), CD86 (B7-2), B7-H1 (CD274), B7-H2 (ICOS Ligand), B7-H3 (CD276), CD28, CTLA-4 (CD152), ICOS, PD-1, PD-L2 (B7-DC),
Catalog No. 50-195-0834