Learn More
Cayman Chemical Arteether, Purity: ≥95%, CAS: 75887-54-6, 25 mg
SDP
Catalog No. 502951715
Encompass_Preferred
An antimalarial compound and derivative of artemisinin; inhibits P. falciparum (IC50 = 1.27 nM in vitro); effective against various strains of P. berghei in mice (ED90 = 0.5-8.2 nM)
Catalog No. 50-295-1715 Supplier Cayman Chemical Supplier No. 1181425 MG
An antimalarial compound and derivative of artemisinin; inhibits P. falciparum (IC50 = 1.27 nM in vitro); effective against various strains of P. berghei in mice (ED90 = 0.5-8.2 nM)