Learn More
Cayman Chemical Arylquin 1, Purity: ≥98%, CAS: 1630743-73-5, 5 mg
SDP
Catalog No. 502966133
Encompass_Preferred
A Par-4 secretagogue; binds to vimentin to displace Par-4 for secretion; induces Par-4 secretion from MEFs and prostate cancer cells in vitro; induces paracrine apoptosis of H1299, HOP92, and H460 cancer cells when co-cultured with Par-4+/+ MEFs
Catalog No. 50-296-6133 Supplier Cayman Chemical Supplier No. 169615 MG
A Par-4 secretagogue; binds to vimentin to displace Par-4 for secretion; induces Par-4 secretion from MEFs and prostate cancer cells in vitro; induces paracrine apoptosis of H1299, HOP92, and H460 cancer cells when co-cultured with Par-4+/+ MEFs