Learn More
Cayman Chemical Aspalatone, Purity: ≥98%, CAS: 147249-33-0, 100 mg
SDP
Catalog No. 502953595
Encompass_Preferred
An anti-platelet aggregator (IC50 = 180 µM, in vitro) that prolongs bleeding time significantly in a rodent model of thromboembolism; at 24 mg/kg, generates antioxidant and neuroprotective effects against kainic acid-induced epilepsy in rat hippocampus
Catalog No. 50-295-3595 Supplier Cayman Chemical Supplier No. 13644100 MG
An anti-platelet aggregator (IC50 = 180 µM, in vitro) that prolongs bleeding time significantly in a rodent model of thromboembolism; at 24 mg/kg, generates antioxidant and neuroprotective effects against kainic acid-induced epilepsy in rat hippocampus