Learn More
Cayman Chemical Aspartame, Purity: ≥98%, CAS: 22839-47-0, 50 g
SDP
Catalog No. 502933323
Encompass_Preferred
A synthetic non-caloric sweetener; metabolized to phenylalanine, aspartic acid, and methanol in the gut; increases plasma ALT and AST activity, induces hepatocyte degeneration and leukocyte infiltration in the liver, and reduces hepatic levels of GSH, GSSG, and γ-GC in mice at 80 mg/kg per day for 90 days
Catalog No. 50-293-3323 Supplier Cayman Chemical Supplier No. 2608950 G
A synthetic non-caloric sweetener; metabolized to phenylalanine, aspartic acid, and methanol in the gut; increases plasma ALT and AST activity, induces hepatocyte degeneration and leukocyte infiltration in the liver, and reduces hepatic levels of GSH, GSSG, and γ-GC in mice at 80 mg/kg per day for 90 days