Learn More
Cayman Chemical AT-130, Purity: ≥98%, CAS: 211364-06-6, 5 mg
SDP
Catalog No. 502909917
Encompass_Preferred
An antiviral agent; a non-nucleoside inhibitor of HBV replication in wild-type and rtL180M, rtM204I, and rtL180M + rtL204V lamivudine-resistant strains of HBV (IC50s = 2.4, 1.6, 5.1, and 1.3 µM, respectively); inhibits viral DNA synthesis in HBV-infected HepAD38 cells (EC50 = 0.13 µM)
Catalog No. 50-290-9917 Supplier Cayman Chemical Supplier No. 354535 MG
An antiviral agent; a non-nucleoside inhibitor of HBV replication in wild-type and rtL180M, rtM204I, and rtL180M + rtL204V lamivudine-resistant strains of HBV (IC50s = 2.4, 1.6, 5.1, and 1.3 µM, respectively); inhibits viral DNA synthesis in HBV-infected HepAD38 cells (EC50 = 0.13 µM)