Learn More
Cayman Chemical (±)-Atenolol, Purity: ≥98%, CAS: 29122-68-7, 5 g
SDP
Catalog No. 502967882
Encompass_Preferred
A β1-AR antagonist (Ki = 1.14 µM); selective for β1-ARs over β2-ARs (Ki = 48.7 µM); delays the onset of hypertension in spontaneously hypertensive rats at 200 mg/kg
Catalog No. 50-296-7882 Supplier Cayman Chemical Supplier No. 172505 G
A β1-AR antagonist (Ki = 1.14 µM); selective for β1-ARs over β2-ARs (Ki = 48.7 µM); delays the onset of hypertension in spontaneously hypertensive rats at 200 mg/kg