Learn More
Cayman Chemical Atosiban, Purity: ≥98%, CAS: 90779-69-4, 10 mg
SDP
Catalog No. 502985211
Encompass_Preferred
A peptide analog of oxytocin that antagonizes the human oxytocin receptor (Ki = 81 nM); has tocolytic actions in the acute phase of imminent preterm birth
Catalog No. 50-298-5211 Supplier Cayman Chemical Supplier No. 2095210 MG
A peptide analog of oxytocin that antagonizes the human oxytocin receptor (Ki = 81 nM); has tocolytic actions in the acute phase of imminent preterm birth