Learn More
Cayman Chemical Atosiban, Purity: ≥98%, CAS: 90779-69-4, 25 mg
SDP
Catalog No. 502985390
Encompass_Preferred
A peptide analog of oxytocin that antagonizes the human oxytocin receptor (Ki = 81 nM); has tocolytic actions in the acute phase of imminent preterm birth
Catalog No. 50-298-5390 Supplier Cayman Chemical Supplier No. 2095225 MG
A peptide analog of oxytocin that antagonizes the human oxytocin receptor (Ki = 81 nM); has tocolytic actions in the acute phase of imminent preterm birth