Learn More
Cayman Chemical ATX-001, Purity: ≥98%, CAS: 1777792-33-2, 5 mg
SDP
Catalog No. 502918592
Encompass_Preferred
An ionizable cationic lipid; has been used in combination with other lipids in the formation of LNPs for the delivery of mRNA in vivo
Catalog No. 50-291-8592 Supplier Cayman Chemical Supplier No. 390375 MG
An ionizable cationic lipid; has been used in combination with other lipids in the formation of LNPs for the delivery of mRNA in vivo