Learn More
Cayman Chemical ANTIBODY ATX-100 25mg
SDP
Catalog No. 502914871
Encompass_Preferred
An ionizable cationic lipid (pKa = 6.38); has been used in the generation of LNPs for the delivery of siRNA; intravenous administration of LNPs containing ATX-100 and encapsulating Factor VII siRNA decrease Factor VII blood levels in mice
Catalog No. 50-291-4871 Supplier Cayman Chemical Supplier No. 3693525 MG
Add to Cart
Add to Cart
An ionizable cationic lipid (pKa = 6.38); has been used in the generation of LNPs for the delivery of siRNA; intravenous administration of LNPs containing ATX-100 and encapsulating Factor VII siRNA decrease Factor VII blood levels in mice