Learn More
Cayman Chemical AUTEN-99, Purity: ≥98%, CAS: 1049780-58-6, 10 mg
SDP
Catalog No. 502989021
Encompass_Preferred
An inhibitor of MRMR14/Jumpy; inhibits MTMR14/Jumpy activity in a cell-free assay at 10 and 100 µM; enhances autophagic flux and increases the levels of LC3B-11 in HeLa cells at 1 and 10 µM; increases survival of mouse primary neurons exposed to hydrogen peroxide as a model of oxidative stress at 0.5-5 µM; increases the number of autophagic structures in mouse pancreas, kidney, and liver; induces autophagy in the Drosophila larval fat body and increases the percentage of flies climbing a glass column in a climbing assay in a transgenic model of Parkinson’s disease
Catalog No. 50-298-9021 Supplier Cayman Chemical Supplier No. 3004710 MG
An inhibitor of MRMR14/Jumpy; inhibits MTMR14/Jumpy activity in a cell-free assay at 10 and 100 µM; enhances autophagic flux and increases the levels of LC3B-11 in HeLa cells at 1 and 10 µM; increases survival of mouse primary neurons exposed to hydrogen peroxide as a model of oxidative stress at 0.5-5 µM; increases the number of autophagic structures in mouse pancreas, kidney, and liver; induces autophagy in the Drosophila larval fat body and increases the percentage of flies climbing a glass column in a climbing assay in a transgenic model of Parkinson’s disease