Learn More
Cayman Chemical AZ 32, Purity: ≥98%, CAS: 2288709-96-4, 10 mg
SDP
Catalog No. 502933396
Encompass_Preferred
An ATM kinase inhibitor (IC50 = 50s = 4.6 and >4.6 µM, respectively); enhances radiation-induced cytotoxicity in a panel of five human glioma cells expressing wild-type or mutant p53; increases survival in a U87-281G glioma orthotopic mouse xenograft model, as well as a NCI H2228 NSCLC mouse xenograft model of metastatic brain tumors, at 200 and 50 mg/kg, respectively, in combination with ionizing radiation
Catalog No. 50-293-3396 Supplier Cayman Chemical Supplier No. 2638010 MG
An ATM kinase inhibitor (IC50 = 50s = 4.6 and >4.6 µM, respectively); enhances radiation-induced cytotoxicity in a panel of five human glioma cells expressing wild-type or mutant p53; increases survival in a U87-281G glioma orthotopic mouse xenograft model, as well as a NCI H2228 NSCLC mouse xenograft model of metastatic brain tumors, at 200 and 50 mg/kg, respectively, in combination with ionizing radiation