Learn More
Cayman Chemical AZ 33, Purity: ≥98%, CAS: 1370290-34-8, 10 mg
SDP
Catalog No. 502935940
Encompass_Preferred
An inhibitor of LDHA (Kd = 0.093 μM; IC50 = 0.5 μM); selective for LDHA over LDHB in an NADH competition assay using recombinant enzymes (IC50s = 0.54 and 3.6 μM, respectively); cell-impermeable and lacks cytotoxicity against HeLa cervical cancer cells (IC50 = >500 μM)
Catalog No. 50-293-5940 Supplier Cayman Chemical Supplier No. 2544410 MG
An inhibitor of LDHA (Kd = 0.093 μM; IC50 = 0.5 μM); selective for LDHA over LDHB in an NADH competition assay using recombinant enzymes (IC50s = 0.54 and 3.6 μM, respectively); cell-impermeable and lacks cytotoxicity against HeLa cervical cancer cells (IC50 = >500 μM)