Learn More
Cayman Chemical AZ82, Purity: ≥98%, CAS: 1449578-65-7, 5 mg
SDP
Catalog No. 502987159
Encompass_Preferred
An inhibitor of KIFC1; binds to the KIFC1/microtubule complex and inhibits its microtubule-stimulated activity (IC50 = 300 nM) in an ATP-competitive (Ki = 43 nM) and microtubule-noncompetitive manner; does not inhibit basal KIFC1 activity at a concentration of 100 µM; selective for KIFC1 over a panel of nine kinesin motor proteins at a concentration of 5 µM; AZ82 preferentially induces multipolar spindle formation in cancer cell lines with high numbers of extra chromosomes
Catalog No. 50-298-7159 Supplier Cayman Chemical Supplier No. 218985 MG
An inhibitor of KIFC1; binds to the KIFC1/microtubule complex and inhibits its microtubule-stimulated activity (IC50 = 300 nM) in an ATP-competitive (Ki = 43 nM) and microtubule-noncompetitive manner; does not inhibit basal KIFC1 activity at a concentration of 100 µM; selective for KIFC1 over a panel of nine kinesin motor proteins at a concentration of 5 µM; AZ82 preferentially induces multipolar spindle formation in cancer cell lines with high numbers of extra chromosomes