Learn More
Cayman Chemical AZD 0095, Purity: ≥98%, CAS: 2750001-23-9, 50 mg
SDP
Catalog No. 502945112
Encompass_Preferred
An MCT4 inhibitor (IC50 = 1.3 nM); selectively inhibits lactate influx in NCI H358 cells (IC50 = 6.1 nM), which highly express MCT4 and do not express MCT1, over lactate influx in K562 cells (IC50 = >30 µM), which highly express MCT1 and do not express MCT4; inhibits the proliferation of NCI H358 cells (IC50 = 26 nM); reduces tumor growth in an NCI H358 mouse xenograft model at 100 mg/kg twice per day in combination with cediranib
Catalog No. 50-294-5112 Supplier Cayman Chemical Supplier No. 4213450 MG
An MCT4 inhibitor (IC50 = 1.3 nM); selectively inhibits lactate influx in NCI H358 cells (IC50 = 6.1 nM), which highly express MCT4 and do not express MCT1, over lactate influx in K562 cells (IC50 = >30 µM), which highly express MCT1 and do not express MCT4; inhibits the proliferation of NCI H358 cells (IC50 = 26 nM); reduces tumor growth in an NCI H358 mouse xenograft model at 100 mg/kg twice per day in combination with cediranib