Learn More
Cayman Chemical AZD 0865, Purity: ≥98%, CAS: 248919-64-4, 1 mg
SDP
Catalog No. 502909741
Encompass_Preferred
An H+/K+-ATPase inhibitor (Ki = 46 nM); inhibits histamine- or dibutyryrl-cAMP-induced acid production in isolated rabbit gastric glands (IC50s = 0.28 and 0.26 µM, respectively)
Catalog No. 50-290-9741 Supplier Cayman Chemical Supplier No. 368051 MG
An H+/K+-ATPase inhibitor (Ki = 46 nM); inhibits histamine- or dibutyryrl-cAMP-induced acid production in isolated rabbit gastric glands (IC50s = 0.28 and 0.26 µM, respectively)