Learn More
Cayman Chemical AZD 2014, Purity: ≥98%, CAS: 1009298-59-2, 10 mg
SDP
Catalog No. 502968287
Encompass_Preferred
A potent, selective, ATP-competitive mTORC1/2 dual inhibitor (IC50 = 2.81 nM against the isolated recombinant enzyme); exhibits >1,000-fold selectivity against all PI3K isoforms; has broad antiproliferative effects across multiple cell lines, reduces tumor growth in in vivo models of ER+ breast cancer, and acts as a radiosensitizer
Catalog No. 50-296-8287 Supplier Cayman Chemical Supplier No. 1737810 MG
A potent, selective, ATP-competitive mTORC1/2 dual inhibitor (IC50 = 2.81 nM against the isolated recombinant enzyme); exhibits >1,000-fold selectivity against all PI3K isoforms; has broad antiproliferative effects across multiple cell lines, reduces tumor growth in in vivo models of ER+ breast cancer, and acts as a radiosensitizer