Learn More
Cayman Chemical AZD 3293, Purity: ≥98%, CAS: 1383982-64-6, 10 mg
SDP
Catalog No. 502935709
Encompass_Preferred
An inhibitor of BACE1 and BACE2 (Kis = 0.4 and 0.8 nM, respectively); selective for BACE1 and BACE2 over cathepsin D, γ-secretase, and hERG (IC50s = 3,797, >25,000, and 4,700 nM, respectively); inhibits the secretion of Aβ40 in primary mouse and guinea pig neuronal cultures (IC50s = 0.61 and 0.31 NM, respectively), as well as in SH-SY5Y cells overexpressing human APP (IC50 = 0.08 nM); reduces plasma and brain levels of Aβ40 and brain levels of Aβ42 in mice at 50, 100, and 200 µmol/kg
Catalog No. 50-293-5709 Supplier Cayman Chemical Supplier No. 2493410 MG
An inhibitor of BACE1 and BACE2 (Kis = 0.4 and 0.8 nM, respectively); selective for BACE1 and BACE2 over cathepsin D, γ-secretase, and hERG (IC50s = 3,797, >25,000, and 4,700 nM, respectively); inhibits the secretion of Aβ40 in primary mouse and guinea pig neuronal cultures (IC50s = 0.61 and 0.31 NM, respectively), as well as in SH-SY5Y cells overexpressing human APP (IC50 = 0.08 nM); reduces plasma and brain levels of Aβ40 and brain levels of Aβ42 in mice at 50, 100, and 200 µmol/kg