Learn More
Cayman Chemical ANTIBODY AZD 5153 10mg
SDP
Catalog No. 502985449
Encompass_Preferred
A bivalent BRD4 inhibitor (IC50 = 5 nM) that simultaneously binds two bromodomains in BRD4; demonstrates antitumor activity in multiple xenograft models of acute myeloid leukemia, multiple myeloma, and diffuse large B cell lymphoma
Catalog No. 50-298-5449 Supplier Cayman Chemical Supplier No. 2086410 MG
A bivalent BRD4 inhibitor (IC50 = 5 nM) that simultaneously binds two bromodomains in BRD4; demonstrates antitumor activity in multiple xenograft models of acute myeloid leukemia, multiple myeloma, and diffuse large B cell lymphoma