Learn More
Cayman Chemical Azilsartan, Purity: ≥98%, CAS: 147403-03-0, 1 g
SDP
Catalog No. 502936153
Encompass_Preferred
An AT1 receptor antagonist (IC50 = 0.42 µM) and the active metabolite of azilsartan medoxomil; acts as an inverse agonist, inhibiting angiotensin II-induced accumulation of inositol-1-phosphate in COS-7 cells expressing recombinant human AT1 (IC50 = 2.6 nM); reduces the maximal contractile response induced by angiotensin II in isolated rabbit aortic strips (pD2 = 9.9); inhibits the angiotensin II-induced pressor response in conscious normotensive rats at 100 ng/kg
Catalog No. 50-293-6153 Supplier Cayman Chemical Supplier No. 260911 G
An AT1 receptor antagonist (IC50 = 0.42 µM) and the active metabolite of azilsartan medoxomil; acts as an inverse agonist, inhibiting angiotensin II-induced accumulation of inositol-1-phosphate in COS-7 cells expressing recombinant human AT1 (IC50 = 2.6 nM); reduces the maximal contractile response induced by angiotensin II in isolated rabbit aortic strips (pD2 = 9.9); inhibits the angiotensin II-induced pressor response in conscious normotensive rats at 100 ng/kg