Learn More
Cayman Chemical BAPTA AM, Purity: ≥98%, CAS: 126150-97-8, 10 mg
SDP
Catalog No. 502973326
Encompass_Preferred
A cell permeable analog of BAPTA that binds calcium only after the AM group is removed by cytoplasmic esterases; commonly used at 10-100 μM to evaluate the role of intracellular calcium in cell signaling; also inhibits Kv channels, including Kv1.3, Kv1.5, and Kv11.1 (Ki = 1.45, 1.23, and 1.30 μM, respectively)
Catalog No. 50-297-3326 Supplier Cayman Chemical Supplier No. 1555110 MG
A cell permeable analog of BAPTA that binds calcium only after the AM group is removed by cytoplasmic esterases; commonly used at 10-100 μM to evaluate the role of intracellular calcium in cell signaling; also inhibits Kv channels, including Kv1.3, Kv1.5, and Kv11.1 (Ki = 1.45, 1.23, and 1.30 μM, respectively)