Learn More
Cayman Chemical BAR501, Purity: ≥95%, CAS: 1632118-69-4, 25 mg
SDP
Catalog No. 502964262
Encompass_Preferred
A selective GP-BAR1 agonist (EC50 = 1 μM); reduces hepatic perfusion pressure and counteracts norepinephrine-induced vasoconstriction in naïve rats; protects against development of endothelial dysfunction by increasing CSE expression and activity and reducing ET-1 gene expression in rat model of cirrhosis; increases Akt-dependent phosphorylation of CSE and eNOS and inhibits ET-1 transcription in vitro; reduces trinitrobenzenesulfonic acid-induced colitis in mice in a dose-dependent manner
Catalog No. 50-296-4262 Supplier Cayman Chemical Supplier No. 2246025 MG
A selective GP-BAR1 agonist (EC50 = 1 μM); reduces hepatic perfusion pressure and counteracts norepinephrine-induced vasoconstriction in naïve rats; protects against development of endothelial dysfunction by increasing CSE expression and activity and reducing ET-1 gene expression in rat model of cirrhosis; increases Akt-dependent phosphorylation of CSE and eNOS and inhibits ET-1 transcription in vitro; reduces trinitrobenzenesulfonic acid-induced colitis in mice in a dose-dependent manner