Learn More
Cayman Chemical Bavachinin, Purity: ≥98%, CAS: 19879-30-2, 5 mg
SDP
Catalog No. 502981864
Encompass_Preferred
A pan-PPAR agonist (EC50s = 4.0, 8.1, and 0.74 µM for PPARα, β/δ, and γ, respectively); has synergistic effects when combined with thiazolidinediones or fibrates; lowers glucose and triacylglycerol levels in db/db mice
Catalog No. 50-298-1864 Supplier Cayman Chemical Supplier No. 193765 MG
A pan-PPAR agonist (EC50s = 4.0, 8.1, and 0.74 µM for PPARα, β/δ, and γ, respectively); has synergistic effects when combined with thiazolidinediones or fibrates; lowers glucose and triacylglycerol levels in db/db mice