Learn More
Cayman Chemical BAY-069, Purity: ≥98%, CAS: 2639638-66-5, 5 mg
SDP
Catalog No. 502944273
Encompass_Preferred
A BCAT1 and BCAT2 inhibitor (IC50s = 31 and 153 nM, respectively); selective for BCAT1 and BCAT2 over a panel of 77 enzymes, receptors, and transporters at 10 µM; increases the levels of BCAAs in U87MG and MDA-MB-231 cancer cells (IC50s = 358 and 874 nM, respectively) but not in the presence of HSA or BSA (IC50s = >50 µM for both)
Catalog No. 50-294-4273 Supplier Cayman Chemical Supplier No. 411495 MG
A BCAT1 and BCAT2 inhibitor (IC50s = 31 and 153 nM, respectively); selective for BCAT1 and BCAT2 over a panel of 77 enzymes, receptors, and transporters at 10 µM; increases the levels of BCAAs in U87MG and MDA-MB-231 cancer cells (IC50s = 358 and 874 nM, respectively) but not in the presence of HSA or BSA (IC50s = >50 µM for both)