Learn More
Cayman Chemical ANTIBODY BAY-179 10mg
SDP
Catalog No. 502913814
Encompass_Preferred
A mitochondrial complex I inhibitor (IC50s = 79, 38, 27, and 47 nM for the human, mouse, rat, and dog enzymes, respectively)
Catalog No. 50-291-3814 Supplier Cayman Chemical Supplier No. 3665710 MG
A mitochondrial complex I inhibitor (IC50s = 79, 38, 27, and 47 nM for the human, mouse, rat, and dog enzymes, respectively)