Learn More
Cayman Chemical BAY-218, Purity: ≥98%, CAS: 2162982-11-6, 10 mg
SDP
Catalog No. 502916260
Encompass_Preferred
An AhR antagonist (IC50 = 39.9 nM); decreases AhR nuclear translocation induced by aflatoxin B1 in L-02 hepatocytes and TK6 lymphoblasts when preincubated at 1 µM; reduces tumor growth in a CT26 mouse xenograft model at 30 mg/kg alone and in combination with an anti-PD-L1 antibody
Catalog No. 50-291-6260 Supplier Cayman Chemical Supplier No. 3758310 MG
An AhR antagonist (IC50 = 39.9 nM); decreases AhR nuclear translocation induced by aflatoxin B1 in L-02 hepatocytes and TK6 lymphoblasts when preincubated at 1 µM; reduces tumor growth in a CT26 mouse xenograft model at 30 mg/kg alone and in combination with an anti-PD-L1 antibody