Learn More
Cayman Chemical BAY-293, Purity: ≥98%, CAS: 2244904-70-7, 25 mg
SDP
Catalog No. 502949239
Encompass_Preferred
An inhibitor of the K-Ras-SOS1 protein-protein interaction (IC50 = 0.021 µM); inhibits proliferation of K562 and MOLM-13 cells expressing wild-type K-RAS as well as NCI H358 and Calu-1 cells expressing mutant K-RASG12C (IC50s = 1.09, 0.995, 3.48, and 3.19 µM, respectively)
Catalog No. 50-294-9239 Supplier Cayman Chemical Supplier No. 3360625 MG
An inhibitor of the K-Ras-SOS1 protein-protein interaction (IC50 = 0.021 µM); inhibits proliferation of K562 and MOLM-13 cells expressing wild-type K-RAS as well as NCI H358 and Calu-1 cells expressing mutant K-RASG12C (IC50s = 1.09, 0.995, 3.48, and 3.19 µM, respectively)