Learn More
Cayman Chemical ANTIBODY BAY-474 25mg
SDP
Catalog No. 502943585
Encompass_Preferred
A c-Met inhibitor (IC50 = <100 nM)
Catalog No. 50-294-3585 Supplier Cayman Chemical Supplier No. 4145125 MG
A c-Met inhibitor (IC50 = <100 nM)