Learn More
Cayman Chemical ANTIBODY BAY-588 5mg
SDP
Catalog No. 502983726
Encompass_Preferred
An inhibitor of glucosylceramide synthase inhibits glucosylceramide synthase by 70 41 and 62 in MDCK cell homogenates mouse liver microsomes and mouse brain homogenates respectively at 20 UM inhibits sphingomyelin synthase activity in P falciparum-infected erythrocytes and inhibits growth of P falciparum (IC50 0 85 UM)
Catalog No. 50-298-3726 Supplier Cayman Chemical Supplier No. 199605 MG
An inhibitor of glucosylceramide synthase inhibits glucosylceramide synthase by 70 41 and 62 in MDCK cell homogenates mouse liver microsomes and mouse brain homogenates respectively at 20 UM inhibits sphingomyelin synthase activity in P falciparum-infected erythrocytes and inhibits growth of P falciparum (IC50 0 85 UM)