Learn More
Cayman Chemical BCI-121, Purity: ≥98%, CAS: 432529-82-3, 50 mg
SDP
Catalog No. 502948225
Encompass_Preferred
An inhibitor of SMYD3; inhibits H3K4 di- and trimethylation in HT-29 cell lysates at 100 µM; inhibits proliferation in a panel of colorectal, lung, pancreatic, prostate, and ovarian cancer cell lines in an SMYD3 expression-dependent manner; induces cell cycle arrest at the S phase and decreases global H5K4 methylation and H3K4 dimethylation in HT-29 cells; intratumoral injection reduces tumor volume in an MHCC97H HCC mouse xenograft model at 20 µM
Catalog No. 50-294-8225 Supplier Cayman Chemical Supplier No. 3385450 MG
An inhibitor of SMYD3; inhibits H3K4 di- and trimethylation in HT-29 cell lysates at 100 µM; inhibits proliferation in a panel of colorectal, lung, pancreatic, prostate, and ovarian cancer cell lines in an SMYD3 expression-dependent manner; induces cell cycle arrest at the S phase and decreases global H5K4 methylation and H3K4 dimethylation in HT-29 cells; intratumoral injection reduces tumor volume in an MHCC97H HCC mouse xenograft model at 20 µM