Learn More
Cayman Chemical BCP-T.A., Purity: ≥98%, CAS: 2786829-70-5, 10 mg
SDP
Catalog No. 502918182
Encompass_Preferred
A ferroptosis inducer; induces ferroptosis in NCI H522 cells an effect that can be blocked by liproxstatin-1; binds to GPX4 and increases the accumulation of lipid peroxides in NCI H522 cells at 0.5 µM; cytotoxic to WI38 cells and MEFs (IC50s = 22 and 10 nM, respectively), as well as NCI H522, HT-1080, MDA-MB-468, MDA-MB-231, and HeLa cancer cells(IC50s = 17, 19, 84, 21, and 242 nM, respectively)
Catalog No. 50-291-8182 Supplier Cayman Chemical Supplier No. 3765910 MG
A ferroptosis inducer; induces ferroptosis in NCI H522 cells an effect that can be blocked by liproxstatin-1; binds to GPX4 and increases the accumulation of lipid peroxides in NCI H522 cells at 0.5 µM; cytotoxic to WI38 cells and MEFs (IC50s = 22 and 10 nM, respectively), as well as NCI H522, HT-1080, MDA-MB-468, MDA-MB-231, and HeLa cancer cells(IC50s = 17, 19, 84, 21, and 242 nM, respectively)