Learn More
Cayman Chemical BDP9066, Purity: ≥98%, CAS: 2226507-04-4, 10 mg
SDP
Catalog No. 502935090
Encompass_Preferred
An inhibitor of MRCKα and MRCKβ (Kis = 0.0136 and 0.0233 nM, respectively); selective for MRCKα and MRCKβ over ROCK1 and ROCK2 (Kis = 18.4 and 5.38 nM, respectively) and a panel of 115 additional kinases at 1 µM; has antiproliferative activity against a panel of 757 cancer cell lines (EC50s = 50 = 64 nM); inhibits the motility of SCC-12 cells; decreases average papilloma volume in a mouse two-stage model of DMBA-initiated and TPA-promoted skin carcinogenesis at 25 µg/animal
Catalog No. 50-293-5090 Supplier Cayman Chemical Supplier No. 2626410 MG
An inhibitor of MRCKα and MRCKβ (Kis = 0.0136 and 0.0233 nM, respectively); selective for MRCKα and MRCKβ over ROCK1 and ROCK2 (Kis = 18.4 and 5.38 nM, respectively) and a panel of 115 additional kinases at 1 µM; has antiproliferative activity against a panel of 757 cancer cell lines (EC50s = 50 = 64 nM); inhibits the motility of SCC-12 cells; decreases average papilloma volume in a mouse two-stage model of DMBA-initiated and TPA-promoted skin carcinogenesis at 25 µg/animal