Learn More
Cayman Chemical Bendazac, Purity: ≥98%, CAS: 20187-55-7, 5 g
SDP
Catalog No. 502912680
Encompass_Preferred
An NSAID; prevents denaturation of lens and serum proteins in vitro; prevents cyanate-induced opacification of the eye lens in a rat model of cataract from 10-40 mM
Catalog No. 50-291-2680 Supplier Cayman Chemical Supplier No. 351275 G
An NSAID; prevents denaturation of lens and serum proteins in vitro; prevents cyanate-induced opacification of the eye lens in a rat model of cataract from 10-40 mM