Learn More
Cayman Chemical Besifovir, Purity: ≥95%, CAS: 441785-25-7, 10 mg
SDP
Catalog No. 502916299
Encompass_Preferred
An antiviral nucleoside derivative; reduces viral replication in Huh7 cells infected with wild-type HBV or a lamivudine-resistant HBV clinical isolate (IC50s = 3.5 and 7.47 µM, respectively); reduces viral replication in Huh7 cells infected with an entecavir-resistant HBV clinical isolate (IC50 = 26 µM)
Catalog No. 50-291-6299 Supplier Cayman Chemical Supplier No. 3757010 MG
An antiviral nucleoside derivative; reduces viral replication in Huh7 cells infected with wild-type HBV or a lamivudine-resistant HBV clinical isolate (IC50s = 3.5 and 7.47 µM, respectively); reduces viral replication in Huh7 cells infected with an entecavir-resistant HBV clinical isolate (IC50 = 26 µM)