Learn More
Cayman Chemical Betaine, Purity: ≥95%, CAS: 107-43-7, 500 g
SDP
Catalog No. 502934740
Encompass_Preferred
A quaternary ammonium compound with diverse biological activities; has roles in osmoregulation and serves as a methyl donor in the conversion of homocysteine to methionine; reduces serum ALT and AST activity, increases serum HDL levels, decreases serum LDL levels, and reduces hepatic steatosis and inflammation in a mouse model of NAFLD induced by a methionine- and choline-deficient diet at 1.5% w/v in drinking water; reduces plasma levels of tHcy, AdoMet, AdoHcy, and cystathionine and attenuates hypercoagulation in a mouse model of homocystinuria
Catalog No. 50-293-4740 Supplier Cayman Chemical Supplier No. 26814500 G
A quaternary ammonium compound with diverse biological activities; has roles in osmoregulation and serves as a methyl donor in the conversion of homocysteine to methionine; reduces serum ALT and AST activity, increases serum HDL levels, decreases serum LDL levels, and reduces hepatic steatosis and inflammation in a mouse model of NAFLD induced by a methionine- and choline-deficient diet at 1.5% w/v in drinking water; reduces plasma levels of tHcy, AdoMet, AdoHcy, and cystathionine and attenuates hypercoagulation in a mouse model of homocystinuria