Learn More
Cayman Chemical BI-1467335, Purity: ≥95%, CAS: 1478364-68-9, 25 mg
SDP
Catalog No. 502989924
Encompass_Preferred
An inhibitor of VAP-1 (IC50 = 50s = >1,000 and >10,000 nM, respectively); reduces plasma total cholesterol, LDL, and glucose levels, as well as decreases the recruitment of macrophages to atherosclerotic lesions and the area of atherosclerotic plaques, in a cholesterol-fed rabbit model at 10 mg/kg per day; reduces K. pneumoniae-induced neutrophil lung infiltration in mice at 6 mg/kg; reduces airway hyperreactivity in a mouse model of rhinovirus-exacerbated allergy-induced asthma
Catalog No. 50-298-9924 Supplier Cayman Chemical Supplier No. 3067225 MG
An inhibitor of VAP-1 (IC50 = 50s = >1,000 and >10,000 nM, respectively); reduces plasma total cholesterol, LDL, and glucose levels, as well as decreases the recruitment of macrophages to atherosclerotic lesions and the area of atherosclerotic plaques, in a cholesterol-fed rabbit model at 10 mg/kg per day; reduces K. pneumoniae-induced neutrophil lung infiltration in mice at 6 mg/kg; reduces airway hyperreactivity in a mouse model of rhinovirus-exacerbated allergy-induced asthma