Learn More
Cayman Chemical BI-3802, Purity: ≥95%, CAS: 2166387-65-9, 1 mg
SDP
Catalog No. 502942871
Encompass_Preferred
A Bcl-6 degrader (DC50 = 20 nM in SU-DHL-4 cells); binds to Bcl-6 in a cell-free assay and inhibits the interaction of Bcl-6 with NCOR1 in a reporter assay (IC50s = ≤3 and 43 nM, respectively); induces polymerization and SIAH1-dependent ubiquitination of Bcl-6 in cell-free assays; enhances imatinib-induced decreases in tumor volume and weight in a GIST-T1 gastrointestinal stromal tumor mouse xenograft model
Catalog No. 50-294-2871 Supplier Cayman Chemical Supplier No. 417311 MG
A Bcl-6 degrader (DC50 = 20 nM in SU-DHL-4 cells); binds to Bcl-6 in a cell-free assay and inhibits the interaction of Bcl-6 with NCOR1 in a reporter assay (IC50s = ≤3 and 43 nM, respectively); induces polymerization and SIAH1-dependent ubiquitination of Bcl-6 in cell-free assays; enhances imatinib-induced decreases in tumor volume and weight in a GIST-T1 gastrointestinal stromal tumor mouse xenograft model