Learn More
Cayman Chemical BI-9627, Purity: ≥95% (mixture of tautomers), CAS: 1204329-34-9, 5 mg
SDP
Catalog No. 502937860
Encompass_Preferred
An NHE1 inhibitor (IC50s = 6 and 31 nM in pHi and human platelet swelling assays, respectively); greater than 30-fold selective for NHE1 over NHE2 and is inactive at NHE3 up to 16 μM in a pHi assay; inhibits phenylephrine-induced hypertrophy in neonatal rat cardiomyocytes; increases recovery of left ventricular developed pressure and inhibits increases in LVEDP in a Langendorff isolated perfused rat heart model of ischemia-reperfusion injury at 100 nM; attenuates decreases in left ventricular end-systolic pressure and increases in LVEDP in coronary artery-ligated rats at 45 and 150 ppm in the diet
Catalog No. 50-293-7860 Supplier Cayman Chemical Supplier No. 276405 MG
An NHE1 inhibitor (IC50s = 6 and 31 nM in pHi and human platelet swelling assays, respectively); greater than 30-fold selective for NHE1 over NHE2 and is inactive at NHE3 up to 16 μM in a pHi assay; inhibits phenylephrine-induced hypertrophy in neonatal rat cardiomyocytes; increases recovery of left ventricular developed pressure and inhibits increases in LVEDP in a Langendorff isolated perfused rat heart model of ischemia-reperfusion injury at 100 nM; attenuates decreases in left ventricular end-systolic pressure and increases in LVEDP in coronary artery-ligated rats at 45 and 150 ppm in the diet