Learn More
Cayman Chemical BI-D1870, Purity: ≥95%, CAS: 501437-28-1, 10 mg
SDP
Catalog No. 502973082
Encompass_Preferred
A cell permeable, ATP-competitive inhibitor of the four vertebrate isoforms of RSK, RSK1-4 (IC50s = 31, 24, 18, and 15 nM, respectively); also significantly inhibits PLK1, Aurora B, MELK, and MST2 at 100 nM
Catalog No. 50-297-3082 Supplier Cayman Chemical Supplier No. 1526410 MG
A cell permeable, ATP-competitive inhibitor of the four vertebrate isoforms of RSK, RSK1-4 (IC50s = 31, 24, 18, and 15 nM, respectively); also significantly inhibits PLK1, Aurora B, MELK, and MST2 at 100 nM