Learn More
Cayman Chemical Bicyclol, Purity: ≥98%, CAS: 118159-48-1, 1 mg
SDP
Catalog No. 502987411
Encompass_Preferred
A hepatoprotective agent; reduces viral DNA and the secretion of HBsAg and HBeAg by 59 and 35%, respectively, in infected 2.2.15 HepG2 cells; reduced DHBV DNA in DHBV-infected ducks at doses ≥0.4 g/kg; reduces the expression of TNF-α and the accumulation of lymphocytes in the liver in a mouse model of concanavalin A-induced liver injury; prevents formation of DEN/PB-induced hepatocellular carcinomas; reduces liver fibrosis in a rat model of bile duct ligation-induced hepatic fibrosis
Catalog No. 50-298-7411 Supplier Cayman Chemical Supplier No. 215511 MG
A hepatoprotective agent; reduces viral DNA and the secretion of HBsAg and HBeAg by 59 and 35%, respectively, in infected 2.2.15 HepG2 cells; reduced DHBV DNA in DHBV-infected ducks at doses ≥0.4 g/kg; reduces the expression of TNF-α and the accumulation of lymphocytes in the liver in a mouse model of concanavalin A-induced liver injury; prevents formation of DEN/PB-induced hepatocellular carcinomas; reduces liver fibrosis in a rat model of bile duct ligation-induced hepatic fibrosis