Learn More
Cayman Chemical Bikinin, Purity: ≥98%, CAS: 188011-69-0, 10 mg
SDP
Catalog No. 502980055
Encompass_Preferred
An inhibitor of several ASK isoforms when given at a concentration of 10 µM, leaving residual kinase activity at less than 10% for ASKα, γ, ε, ζ, η, ι, and 20% for ASKθ, without inhibiting isoforms β and δ; activates signaling induced by brassinosteroids
Catalog No. 50-298-0055 Supplier Cayman Chemical Supplier No. 1838110 MG
An inhibitor of several ASK isoforms when given at a concentration of 10 µM, leaving residual kinase activity at less than 10% for ASKα, γ, ε, ζ, η, ι, and 20% for ASKθ, without inhibiting isoforms β and δ; activates signaling induced by brassinosteroids